missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Immobilon™ Western Chemiluminescent HRP Substrate (ECL)
Provides high sensitivity over a broad detection range at an economical price
$95.70 - $502.00
Specifications
For Use With (Application) | Dot/Slot Blot |
---|---|
Substrate | Horseradish Peroxidase |
Content And Storage | 2°C to 8°C |
Includes: Luminol, peroxide solution
Catalog Number | Mfr. No. | Kit Contents | Quantity | Sufficient For | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Contents | Quantity | Sufficient For | Price | Quantity | |||
WBKLS0050
|
MilliporeSigma™
WBKLS0050 |
25mL luminol, 25mL peroxide solution | 2 x 25 mL | 500cm2 Membrane |
Each for $95.70
|
|
|||
WBKLS0500
|
MilliporeSigma™
WBKLS0500 |
250mL luminol, 250mL peroxide solution | 2 x 250 mL | 5000cm2 Membrane |
Each for $502.00
|
|
|||
WBKLS0100
|
MilliporeSigma™
WBKLS0100 |
50mL luminol, 50mL peroxide solution | 2 x 50 mL | 1000cm2 Membrane |
Each for $168.00
|
|
|||
Description
- Combines high sensitivity and low background
- Allows detection of high- and low-abundance proteins in the mid-femtogram range
- Lowers antibody consumption by two- to five-fold compared to less sensitive reagents
- Compatible with PVDF and nitrocellulose membranes as well as all commonly used buffers and blocking reagents
Specifications
Dot/Slot Blot | |
Horseradish Peroxidase | |
2°C to 8°C |